accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFFFFFHOFFFFFF

CAS RN 918-21-8

IUPAC Name: 1,1,1,4,4,4-hexafluoro-2,3-bis(trifluoromethyl)butane-2,3-diol
Synonyms: 1,1,1,4,4,4-hexafluoro-2,3-bis(trifluoromethyl)butane-2,3-diol
Molecular Weight (g/mol): 334.06
Molecular Formula: C6H2F12O2
InChi Key: GKDCWKGUOZVDFX-UHFFFAOYSA-N
SMILES: OC(C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)