Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol 98.0+%, TCI America™
SDP

Catalog No. H127925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H127925G 25 g
H12795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H127925G Supplier TCI America Supplier No. H127925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 918-21-8
Molecular Formula C6H2F12O2
Molecular Weight (g/mol) 334.06
MDL Number MFCD00427700
InChI Key GKDCWKGUOZVDFX-UHFFFAOYSA-N
Synonym Perfluoropinacol
PubChem CID 13518
IUPAC Name 1,1,1,4,4,4-hexafluoro-2,3-bis(trifluoromethyl)butane-2,3-diol
SMILES OC(C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 26°C
Color White-Yellow
Boiling Point 129°C
Quantity 25 g
Formula Weight 334.06
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol
RTECSNumber EK0650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.