accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HOH2C

CAS RN 918-85-4

IUPAC Name: 3-methylpent-1-en-3-ol
Synonyms: 1-penten-3-ol, 3-methyl-
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: HFYAEUXHCMTPOL-UHFFFAOYNA-N
SMILES: CCC(C)(O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results