Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-1-penten-3-ol, 98%, Thermo Scientific™

Catalog No. AAB2089306
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2089306 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2089306 Supplier Thermo Scientific Chemicals Supplier No. B2089306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed in the optimized palladium catalyst systems for the selective liquid-phase hydrogenation of functionalized alkynes and in the bacterial degradation of tert-amyl alcohol proceeds via hemiterpene 2-methyl-3-buten-2-ol by employing the tertiary alcohol desaturase function of the rieske nonheme mononuclear iron oxygenase MdpJ.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Specifications

CAS 918-85-4
Boiling Point 118°C
Flash Point 26°C (79°F)
Molecular Formula C6H12O
Quantity 5g
MDL Number MFCD00004481
UN Number UN1987
Synonym 3-methyl-1-penten-3-ol, ethylbutenol, 1-penten-3-ol, 3-methyl, methylethylvinylcarbinol, 3-methyl-penten-1-ol-3, 3-methyl-penten-1-ol-3 german, 4-01-00-02147 beilstein handbook reference, .+/-.-3-methyl-1-penten-3-ol, +/--3-methyl-1-penten-3-ol
InChI Key HFYAEUXHCMTPOL-UHFFFAOYNA-N
SMILES CCC(C)(O)C=C
IUPAC Name 3-methylpent-1-en-3-ol
Molecular Weight (g/mol) 100.16
PubChem CID 13519
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 3-Methyl-1-penten-3-ol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECSNumber SB3495000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.