accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 918311-87-2

IUPAC Name: [(pyridin-3-yl)methyl](triphenylmethyl)amine
Synonyms: (pyridin-3-ylmethyl)(triphenylmethyl)amine
Molecular Weight (g/mol): 350.47
Molecular Formula: C25H22N2
InChi Key: PQFNWDHABGBCHB-UHFFFAOYSA-N
SMILES: C(NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results