CAS RN 918870-43-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific Chemicals Lck Inhibitor II
CAS: 918870-43-6 Molecular Formula: C24H26N6O3 Synonym: 3-(2-(1H-Benzo[d]imidazol-1-yl)-6-(2-morpholinoethoxy)-pyrimidin-4-ylamino)-4-methylphenol;