accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 919-19-7

IUPAC Name: diethyl acetylphosphonate
Synonyms: diethyl acetylphosphonate
Molecular Weight (g/mol): 180.14
Molecular Formula: C6H13O4P
InChi Key: YOHJPFQGGNEGSE-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results