Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Acetylphosphonate 97.0+%, TCI America™
SDP

Catalog No. A148025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A148025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A148025G Supplier TCI America Supplier No. A148025G

missing translation for 'validMainframeMsg'

Specifications

CAS 919-19-7
Color Yellow
Boiling Point 115°C
Molecular Formula C6H13O4P
MDL Number MFCD00015138
Quantity 25 g
Synonym Acetylphosphonic Acid Diethyl Ester
InChI Key YOHJPFQGGNEGSE-UHFFFAOYSA-N
SMILES CCOP(=O)(C(=O)C)OCC
IUPAC Name 1-diethoxyphosphorylethanone
Molecular Weight (g/mol) 180.14
PubChem CID 522997
Formula Weight 180.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Acetylphosphonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.