accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 919-29-9

IUPAC Name: 1,1,1-triethoxypentane
Synonyms: 1,1,1-triethoxypentane
Molecular Weight (g/mol): 204.31
Molecular Formula: C11H24O3
InChi Key: DIKAUBKIDNXNNW-UHFFFAOYSA-N
SMILES: CCCCC(OCC)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results