accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-00-2

IUPAC Name: 2-[(3-chlorophenyl)(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: 2-[(3-chlorophenyl)(2-hydroxyethyl)amino]ethanol
Molecular Weight (g/mol): 215.68
Molecular Formula: C10H14ClNO2
InChi Key: MVQUJEUCFOGFJU-UHFFFAOYSA-N
SMILES: OCCN(CCO)C1=CC=CC(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results