Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(2-hydroxyethyl)-3-chloroaniline 90.0+%, TCI America™
SDP

Catalog No. B049025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B049025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B049025G Supplier TCI America Supplier No. B049025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92-00-2
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00002852
InChI Key MVQUJEUCFOGFJU-UHFFFAOYSA-N
Synonym 3-Chloro-N,N-di(2-hydroxyethyl)aniline, N-(3-Chlorophenyl)diethanolamine, 2,2′C-[(3-Chlorophenyl)imino]diethanol, N,N-Di(2-hydroxyethyl)-3-chloroaniline
PubChem CID 66688
IUPAC Name 2-[(3-chlorophenyl)(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)C1=CC=CC(Cl)=C1

Specifications

Melting Point 84°C
Quantity 25 g
Formula Weight 215.68
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(2-hydroxyethyl)-3-chloroaniline
EINECSNumber (3)-0227
RTECSNumber KK2420000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.