accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92-06-8

IUPAC Name: 3-phenyl-1,1'-biphenyl
Synonyms: m-terphenyl
Molecular Weight (g/mol): 230.31
Molecular Formula: C18H14
InChi Key: YJTKZCDBKVTVBY-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC(=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Boiling Point 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results