accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 92-82-0

IUPAC Name: phenazine
Synonyms: phenazine acridizine azophenylene dibenzoparadiazine dibenzopyrazine dibenzo-p-diazine
Molecular Weight (g/mol): 180.21
Molecular Formula: C12H8N2
InChi Key: PCNDJXKNXGMECE-UHFFFAOYSA-N
SMILES: C1=CC2=NC3=CC=CC=C3N=C2C=C1
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (7)
  • (3)
  • (1)
Percent Purity 
  • (4)
  • (3)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (7)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)