Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenazine, 99+%, Thermo Scientific™

Catalog No. AA4214204
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4214204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4214204 Supplier Thermo Scientific Chemicals Supplier No. 4214204

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-82-0
Molecular Formula C12H8N2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00005023
InChI Key PCNDJXKNXGMECE-UHFFFAOYSA-N
Synonym dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
PubChem CID 4757
ChEBI CHEBI:36674
IUPAC Name phenazine
SMILES C1=CC=C2C(=C1)N=C3C=CC=CC3=N2

Specifications

Melting Point 175°C to 177°C
Quantity 2 g
Merck Index 14,7217
Solubility Information Soluble in mineral acids, moderately soluble in ether and benzene; Slightly soluble in alcohol
Formula Weight 180.21
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Phenazine
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H340
Toxic if swallowed.
May cause genetic defects.
Precautionary Statement GHS P Statement
P281-P301+P310-P308+P313-P321-P405-P501a
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF exposed or concerned: Get medical advice/attention.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.