accessibility menu, dialog, popup

UserName

Viewing profile as user:
SS

CAS RN 92-85-3

IUPAC Name: thianthrene
Synonyms: thianthrene
Molecular Weight (g/mol): 216.32
Molecular Formula: C12H8S2
InChi Key: GVIJJXMXTUZIOD-UHFFFAOYSA-N
SMILES: S1C2=CC=CC=C2SC2=CC=CC=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)