accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92022-50-9

IUPAC Name: 1-[(benzenesulfonyl)methyl]-2-bromobenzene
Synonyms: 1-[(benzenesulfonyl)methyl]-2-bromobenzene
Molecular Weight (g/mol): 311.19
Molecular Formula: C13H11BrO2S
InChi Key: CQNGEPLMWCLSBR-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1CS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results