accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CCH3NNCCH3H3CCH3CH3OHOCu(E/Z)

CAS RN 920739-11-3

IUPAC Name: 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper
Synonyms: 1,3-bis(2,6-diisopropylphenyl)-2H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper
Molecular Weight (g/mol): 676.4
Molecular Formula: C42H48CuN2O2
InChi Key: QEXNUQSCWMWURE-UHFFFAOYSA-N
SMILES: [Cu].OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results