Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I) 98.0+%, TCI America™
SDP

Catalog No. B3351200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3351200MG 200 mg
B33511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3351200MG Supplier TCI America Supplier No. B3351200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 920739-11-3
Molecular Formula C42H48CuN2O2
Molecular Weight (g/mol) 676.40
MDL Number MFCD12545959
InChI Key QEXNUQSCWMWURE-UHFFFAOYSA-N
PubChem CID 91659139
IUPAC Name 1,3-bis[2,6-bis(propan-2-yl)phenyl]-2,3-dihydro-1H-imidazol-2-ylidene 3-hydroxy-1,3-diphenylprop-2-en-1-one copper
SMILES [Cu].OC(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.CC(C)C1=CC=CC(C(C)C)=C1N1[C]N(C=C1)C1=C(C=CC=C1C(C)C)C(C)C

Specifications

Quantity 200 mg
Formula Weight 675.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene](1,3-diphenyl-1,3-propanedionato)copper(I)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.