CAS RN 920966-17-2
IUPAC Name:
tert-butyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Synonyms:
tert-butyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Molecular Weight (g/mol):
229.32
Molecular Formula:
C12H23NO3
InChi Key:
LBJSEPNOVVUVJA-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC1CCCC(CO)C1
1
–
1
of
1
results
1
Promotions Available
trans-3-(Boc-amino)cyclohexanemethanol, 97%
trans-3-(Boc-amino)cyclohexanemethanol, 97%
CAS: 920966-17-2 Molecular Formula: C12H23NO3 Molecular Weight (g/mol): 229.32 MDL Number: MFCD03844607 InChI Key: LBJSEPNOVVUVJA-NXEZZACHSA-N Synonym: tert-butyl trans-3-hydroxymethyl-cyclohexylcarbamate,tert-butyl trans-3-hydroxymethyl cyclohexylcarbamate,tert-butyl trans-3-hydroxymethylcyclohexyl carbamate,tert-butyl trans-3-hydroxymethyl cyclohexyl carbamate,tert-butyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate PubChem CID: 24720931 IUPAC Name: tert-butyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate SMILES: CC(C)(C)OC(=O)NC1CCCC(C1)CO