Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-(Boc-amino)cyclohexanemethanol, 97%

Catalog No. AAH62934MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62934MD 250 mg
AAH6293403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62934MD Supplier Thermo Scientific Chemicals Supplier No. H62934MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 920966-17-2 | C12H23NO3 | 229.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920966-17-2
Molecular Formula C12H23NO3
Molecular Weight (g/mol) 229.32
MDL Number MFCD03844607
InChI Key LBJSEPNOVVUVJA-NXEZZACHSA-N
Synonym tert-butyl trans-3-hydroxymethyl-cyclohexylcarbamate, tert-butyl trans-3-hydroxymethyl cyclohexylcarbamate, tert-butyl trans-3-hydroxymethylcyclohexyl carbamate, tert-butyl trans-3-hydroxymethyl cyclohexyl carbamate, tert-butyl n-1r,3r-3-hydroxymethyl cyclohexyl carbamate
PubChem CID 24720931
IUPAC Name tert-butyl N-[(1R,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)CO

Specifications

Quantity 250 mg
Formula Weight 229.31
Percent Purity 97%
Chemical Name or Material trans-3-(Boc-amino)cyclohexanemethanol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.