accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OH2NHCl

CAS RN 920966-29-6

IUPAC Name: 3-aminocyclohexane-1-carboxamide hydrochloride
Synonyms: 3-aminocyclohexane-1-carboxamide hydrochloride
Molecular Weight (g/mol): 178.66
Molecular Formula: C7H15ClN2O
InChi Key: ZCFBGXVDLNNMDE-UHFFFAOYNA-N
SMILES: Cl.NC1CCCC(C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)