accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHOHHOHO

CAS RN 921-60-8

IUPAC Name: 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: galactose d-galactose glucose oxidase mannose D-hexose D-hexoses
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: WQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILES: OCC1OC(O)C(O)C(O)C1O
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (7)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (7)
  • (3)