accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONHNCH3

CAS RN 921938-59-2

IUPAC Name: methyl({[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methyl})amine
Synonyms: methyl({[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: FWMVARCHFJQGJP-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(=C1)C1=NC(C)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results