accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOCH2H2C

CAS RN 922-65-6

IUPAC Name: penta-1,4-dien-3-ol
Synonyms: 1,4-pentadien-3-ol
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: ICMWSAALRSINTC-UHFFFAOYSA-N
SMILES: OC(C=C)C=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)