Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Pentadien-3-ol

Catalog No. 3246635G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-466-35G 5 g
32-466-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-466-35G Supplier Sigma Aldrich Supplier No. 3246635G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful building block in natural product synthesis.

Specifications

CAS 922-65-6
Density 0.865 g/mL (at 25°C (literature))
Boiling Point 115°C to 116°C (lit.)
Molecular Formula C5H8O
Refractive Index n20/D 1.445 (literature)
Linear Formula CH2=CHCH(OH)CH=CH2
MDL Number MFCD00008633
Quantity 5 g
Synonym Divinyl carbinol
Molecular Weight (g/mol) 84.12
Percent Purity ≥96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.