CAS RN 92276-38-5
IUPAC Name:
6-bromo-2H-[1,2,3]triazolo[4,5-b]pyridine
Synonyms:
6-bromo-2H-[1,2,3]triazolo[4,5-b]pyridine
Molecular Weight (g/mol):
199.01
Molecular Formula:
C5H3BrN4
InChi Key:
YKWWQNVTTSHHGH-UHFFFAOYSA-N
SMILES:
BrC1=CC2=NNN=C2N=C1
1
–
1
of
1
results
1
6-Bromo-1H-1,2,3-triazolo[4,5-b]pyridine 98.0+%, TCI America™
CAS: 92276-38-5 Molecular Formula: C5H3BrN4 Molecular Weight (g/mol): 199.011 InChI Key: YKWWQNVTTSHHGH-UHFFFAOYSA-N Synonym: 6-Bromo-4-azabenzotriazole PubChem CID: 494193 IUPAC Name: 6-bromo-2H-triazolo[4,5-b]pyridine SMILES: C1=C(C=NC2=NNN=C21)Br