Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-1H-1,2,3-triazolo[4,5-b]pyridine 98.0+%, TCI America™
SDP

Catalog No. B30341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B30341G Supplier TCI America Supplier No. B30341G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92276-38-5
Molecular Formula C5H3BrN4
Molecular Weight (g/mol) 199.011
InChI Key YKWWQNVTTSHHGH-UHFFFAOYSA-N
Synonym 6-Bromo-4-azabenzotriazole
PubChem CID 494193
IUPAC Name 6-bromo-2H-triazolo[4,5-b]pyridine
SMILES C1=C(C=NC2=NNN=C21)Br

Specifications

Melting Point 206°C
Color Yellow
Quantity 1 g
Formula Weight 199.01
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-1H-1,2,3-triazolo[4,5-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.