accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBrOHO

CAS RN 923-06-8

IUPAC Name: 2-bromobutanedioic acid
Synonyms: bromosuccinic acid
Molecular Weight (g/mol): 196.98
Molecular Formula: C4H5BrO4
InChi Key: QQWGVQWAEANRTK-UHFFFAOYNA-N
SMILES: OC(=O)CC(Br)C(O)=O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)