Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Bromosuccinic acid, 96%, Thermo Scientific™

Catalog No. AC107441000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC107441000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC107441000 Supplier Thermo Scientific Chemicals Supplier No. 107441000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 923-06-8
Density 2.0730g/mL
Assay Percent Range 96%
Molecular Formula C4H5BrO4
Linear Formula HO2CCH2CH(Br)CO2H
MDL Number MFCD00004213
Quantity 100g
Beilstein 02, 621
Merck Index 15, 1447
Synonym bromosuccinic acid, 2-bromosuccinic acid, monobromosuccinic acid, butanedioic acid, bromo, +--bromosuccinic acid, dl-bromosuccinic acid, rs-bromosuccinic acid, succinic acid, bromo, bromobutanedioic acid
InChI Key QQWGVQWAEANRTK-UHFFFAOYNA-N
SMILES OC(=O)CC(Br)C(O)=O
IUPAC Name 2-bromobutanedioic acid
Molecular Weight (g/mol) 196.98
PubChem CID 73557
ChEBI CHEBI:73712
Formula Weight 196.99
Chemical Name or Material DL-Bromosuccinic acid, 96%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.