accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOO

CAS RN 92343-46-9

IUPAC Name: 2-hydroxy-4-oxatricyclo[4.2.1.0³,⁷]nonan-5-one
Synonyms: 2-hydroxy-4-oxatricyclo[4.2.1.0³,⁷]nonan-5-one
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: SCDJKGVPVLZLED-UHFFFAOYNA-N
SMILES: OC1C2CC3C(C2)C(=O)OC13
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results