accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNBrHCl

CAS RN 923590-95-8

IUPAC Name: 4-bromo-2,3-dihydro-1H-isoindole hydrochloride
Synonyms: 4-bromo-2,3-dihydro-1H-isoindole hydrochloride
Molecular Weight (g/mol): 234.52
Molecular Formula: C8H9BrClN
InChi Key: FQHLHVFOJBANKY-UHFFFAOYSA-N
SMILES: Cl.BrC1=CC=CC2=C1CNC2
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)