Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromoisoindoline hydrochloride, 97%

Catalog No. AC435870050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC435870050 5 g Glass bottle
AC435870010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435870050 Supplier Thermo Scientific Chemicals Supplier No. 435870050
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 923590-95-8 | C8H8BrN·ClH | 234.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 923590-95-8
Molecular Formula C8H8BrN·ClH
Molecular Weight (g/mol) 234.52
InChI Key FQHLHVFOJBANKY-UHFFFAOYSA-N
Synonym 4-bromoisoindoline hydrochloride, 4-bromo-isoindoline hcl, 4-bromo-2,3-dihydro-1h-isoindole hydrochloride, 4-bromo-1h-isoindoline hydrochloride, 4-bromoisoindoline hcl, 1h-isoindole, 4-bromo-2,3-dihydro-, hydrochloride 1:1, pubchem14849, acmc-209y8q, 4-bromo-isoindoline hydrochloride
PubChem CID 44228644
IUPAC Name 4-bromo-2,3-dihydro-1H-isoindole;hydrochloride
SMILES C1C2=C(CN1)C(=CC=C2)Br.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 269°C to 274°C
Color White to Yellow
Elemental Analysis 39.33 to 42.61 % C 5.73 to 6.21 % N
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 234.52
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoisoindoline hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.