accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNNH2

CAS RN 92394-00-8

IUPAC Name: 1-[4-(4-aminophenyl)piperazin-1-yl]ethan-1-one
Synonyms: 1-[4-(4-aminophenyl)piperazin-1-yl]ethanone
Molecular Weight (g/mol): 219.29
Molecular Formula: C12H17N3O
InChi Key: AFVUJJNEILZYJQ-UHFFFAOYSA-N
SMILES: CC(=O)N1CCN(CC1)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results