Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetyl-4-(4-aminophenyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAH2648606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2648606 5 g
AAH2648603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2648606 Supplier Thermo Scientific Chemicals Supplier No. H2648606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92394-00-8
Molecular Formula C12H17N3O
Molecular Weight (g/mol) 219.288
MDL Number MFCD01365904
InChI Key AFVUJJNEILZYJQ-UHFFFAOYSA-N
Synonym 1-4-4-aminophenyl piperazin-1-yl ethanone, 1-acetyl-4-4-aminophenyl piperazine, 4-4-acetyl-1-piperazinyl aniline, 1-4-4-amino-phenyl-piperazin-1-yl-ethanone, 4-4-acetylpiperazin-1-yl aniline, 4-4-acetyl-piperazin-1-yl aniline, 1-4-4-aminophenyl piperazin-1-yl ethan-1-one, 4-4-acetylpiperazin-1-yl phenyl amine
PubChem CID 736269
IUPAC Name 1-[4-(4-aminophenyl)piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)N

Specifications

Quantity 5 g
Formula Weight 219.29
Percent Purity 97%
Chemical Name or Material 1-Acetyl-4-(4-aminophenyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.