accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONCH3H3C(Z)

CAS RN 924-99-2

IUPAC Name: ethyl (2Z)-3-(dimethylamino)prop-2-enoate
Synonyms: ethyl (2Z)-3-(dimethylamino)prop-2-enoate
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: MVUMJYQUKKUOHO-WAYWQWQTSA-N
SMILES: CCOC(=O)\C=C/N(C)C
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)