Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-(N,N-dimethylamino)acrylate, 99+%

Catalog No. AC269631000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC269631000 100 g Glass bottle
AC269630250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC269631000 Supplier Thermo Scientific Chemicals Supplier No. 269631000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 924-99-2 | C7H13NO2 | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-99-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00144269
InChI Key MVUMJYQUKKUOHO-AATRIKPKSA-N
Synonym ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
PubChem CID 5369162
IUPAC Name ethyl (E)-3-(dimethylamino)prop-2-enoate
SMILES CCOC(=O)C=CN(C)C

Specifications

Melting Point 17.0°C to 18.0°C
Color Orange to Yellow
Density 1.0000g/mL
Boiling Point 118°C to 121°C (10.0 HPA)
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH=CHCOOC2H5
Refractive Index 1.5105 to 1.5125
Quantity 100 g
Specific Gravity 1
Solubility Information Solubility in water: 90g/L (25 c)
Formula Weight 143.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl 3-(N, N-dimethylamino)acrylate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.