accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOSOOHNH2NHCl

CAS RN 92504-03-5

IUPAC Name: N-(2-aminoethyl)-2-nitrobenzene-1-sulfonamide hydrochloride
Synonyms: N-(2-aminoethyl)-2-nitrobenzenesulfonamide hydrochloride
Molecular Weight (g/mol): 281.71
Molecular Formula: C8H12ClN3O4S
InChi Key: ZEWOSYWRSUZSQX-UHFFFAOYSA-N
SMILES: Cl.NCCNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results