Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Aminoethyl)-2-nitrobenzenesulfonamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A23291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23291G Supplier TCI America Supplier No. A23291G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 92504-03-5
Molecular Formula C8H12ClN3O4S
Quantity 1 g
MDL Number MFCD07366919
Synonym 1-Amino-2-(2-nitrobenzenesulfonamido)ethane Hydrochloride, N-(2-Nitrobenzenesulfonyl)ethylenediamine Hydrochloride, N-(o-Ns)-1,2-diaminoethane Hydrochloride
InChI Key ZEWOSYWRSUZSQX-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)NCCN.Cl
IUPAC Name N-(2-aminoethyl)-2-nitrobenzenesulfonamide;hydrochloride
Molecular Weight (g/mol) 281.711
PubChem CID 53384390
Formula Weight 281.71
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Aminoethyl)-2-nitrobenzenesulfonamide Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.