accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNH2

CAS RN 92511-32-5

IUPAC Name: 3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Synonyms: 3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: FCYFJGGJCJDCPB-UHFFFAOYNA-N
SMILES: NC1C2CC(C=C2)C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results