Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%

Catalog No. AC265690010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC265690010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC265690010 Supplier Thermo Scientific Chemicals Supplier No. 265690010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 92511-32-5 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 261.0°C to 263.0°C
CAS 92511-32-5
CAS Min % 97.5
CAS Max % 100.0
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C8H11NO2
MDL Number MFCD00143158
Synonym diendo-3-amino-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, rac-1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-endo-aminobicyclo 2.2.1 hept-5-ene-2-endo-carboxylic acid, 1r,2r,3s,4s/1s,2s,3r,4r-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 1r,1beta,4s,4beta-3alpha-amino-2alpha-norborna-5-enecarboxylic acid, 1r,2s,3 r,4s-3-amino-5-bicyclo 2,2,1 heptene-2-carboxylic acid, 1r,2s,3r,4s-3-aminobicyclo 2.2.1 hept-5-ene-2-carboxylic acid, bicyclo 2.2.1 hept-5-ene-2-carboxylicacid, 3-amino-, 1r,2s,3r,4s, bicyclo 2.2.1 hept-5-ene-2-carboxylic acid, 3-amino-, 1r,2s,3r,4s-9ci
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and methanol
InChI Key FCYFJGGJCJDCPB-BNHYGAARSA-N
SMILES C1C2C=CC1C(C2C(=O)O)N
IUPAC Name (1R,2S,3R,4S)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Quantity 1 g
Molecular Weight (g/mol) 153.18
PubChem CID 13160880
Formula Weight 153.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.