accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 925252-30-8

IUPAC Name: 2-(4-fluorophenyl)-N'-hydroxyethanimidamide
Synonyms: 2-(4-fluorophenyl)-N'-hydroxyethanimidamide
Molecular Weight (g/mol): 168.17
Molecular Formula: C8H9FN2O
InChi Key: VVFOLYZJDVRHLW-UHFFFAOYSA-N
SMILES: NC(CC1=CC=C(F)C=C1)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results