accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 925698-75-5

CH3NHOH2N(E/Z)

CAS RN 925698-75-5

IUPAC Name: N'-hydroxy-2-phenylpropanimidamide
Synonyms: N'-hydroxy-2-phenylpropanimidamide
Molecular Weight (g/mol): 164.21
Molecular Formula: C9H12N2O
InChi Key: KFRRAHGTTHZCNG-UHFFFAOYNA-N
SMILES: CC(C(N)=NO)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results