accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3

CAS RN 926-82-9

IUPAC Name: 3,5-dimethylheptane
Synonyms: heptane, 3,5-dimethyl-
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: DZJTZGHZAWTWGA-UHFFFAOYNA-N
SMILES: CCC(C)CC(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results