Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylheptane 99.0+%, TCI America™
SDP

Catalog No. D120805ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D120805ML 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D120805ML Supplier TCI America Supplier No. D12080.5ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 926-82-9
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
MDL Number MFCD00048659
InChI Key DZJTZGHZAWTWGA-UHFFFAOYNA-N
PubChem CID 13558
IUPAC Name 3,5-dimethylheptane
SMILES CCC(C)CC(C)CC

Specifications

Color Colorless
Boiling Point 136°C
Quantity 0.5 mL
UN Number 1920
Formula Weight 128.26
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylheptane
EINECSNumber (2)-0009
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.