accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(R)ClOClO

CAS RN 92600-11-8

IUPAC Name: (1R)-1-chloro-2-methylpropyl carbonochloridate
Synonyms: (1R)-1-chloro-2-methylpropyl carbonochloridate
Molecular Weight (g/mol): 171.02
Molecular Formula: C5H8Cl2O2
InChi Key: BOGLZTPABVHWDD-BYPYZUCNSA-N
SMILES: CC(C)[C@@H](Cl)OC(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results