accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 92665-29-7

IUPAC Name: (6R,7R)-7-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-8-oxo-3-[(1E)-prop-1-en-1-yl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cefprozil serozil cefprozil anhydrous arzimol procef cefprozilo [inn-spanish] cronocef brisoral
Molecular Weight (g/mol): 389.43
Molecular Formula: C18H19N3O5S
InChi Key: WDLWHQDACQUCJR-ZAMMOSSLSA-N
SMILES: C\C=C\C1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=C(O)C=C1)C2=O)C(O)=O
Color 
  • (1)
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results