accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 927-77-5

MgCH2CH3Br

CAS RN 927-77-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 147.3
Molecular Formula: C3H7BrMg
InChi Key: UGVPKMAWLOMPRS-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC[CH2-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results