Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylmagnesium Bromide (ca. 27% in Tetrahydrofuran, ca. 2mol/L), TCI America™
SDP

Catalog No. P0880250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0880250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0880250G Supplier TCI America Supplier No. P0880250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 927-77-5
Molecular Formula C3H7BrMg
Molecular Weight (g/mol) 147.298
MDL Number MFCD00058928
InChI Key UGVPKMAWLOMPRS-UHFFFAOYSA-M
PubChem CID 101916
IUPAC Name magnesium;propane;bromide
SMILES CC[CH2-].[Mg+2].[Br-]

Specifications

Quantity 250 g
UN Number 3399
Formula Weight 147.30
Chemical Name or Material Propylmagnesium Bromide (ca. 27% in Tetrahydrofuran, ca. 2mol/L)
EINECSNumber (2)-3468
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.