accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 927-86-6

IUPAC Name: [2-(acetyloxy)ethyl]trimethylazanium perchlorate
Synonyms: acetylcholine perchlorate
Molecular Weight (g/mol): 245.66
Molecular Formula: C7H16ClNO6
InChi Key: SHIQLFRCVFUYEK-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CC(=O)OCC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results