Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcholine Perchlorate, TCI America™
SDP

Catalog No. A099810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A099810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A099810G Supplier TCI America Supplier No. A099810G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 927-86-6
Molecular Formula C7H16ClNO6
Molecular Weight (g/mol) 245.656
MDL Number MFCD00036334
InChI Key SHIQLFRCVFUYEK-UHFFFAOYSA-M
PubChem CID 93565
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;perchlorate
SMILES CC(=O)OCC[N+](C)(C)C.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 115°C
Color White
Quantity 10 g
UN Number 1479
Formula Weight 245.66
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Acetylcholine Perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.